Assay Details
| Gene Symbol: kdelr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011046.1 | NP_001011046.1 | 1 - 2 | 195 | 74 | |
| GenBank mRNA | BC084170.1 | - | 1 - 2 | 195 | 74 | |
| CR761657.2 | - | 1 - 2 | 225 | 74 | ||
More Information
Set Membership: |
|

kdelr2
Genomic Map