Assay Details
| Gene Symbol: timm23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011053.1 | NP_001011053.1 | 5 - 6 | 458 | 84 | |
| XM_018095361.1 | XP_017950850.1 | 5 - 6 | 396 | 84 | ||
| GenBank mRNA | BC084178.1 | - | 5 - 6 | 458 | 84 | |
| CT485778.1 | - | 5 - 6 | 469 | 84 | ||
More Information
Set Membership: |
|

timm23
Genomic Map