Assay Details
| Gene Symbol: timm23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011053.1 | NP_001011053.1 | 6 - 7 | 570 | 83 | |
| GenBank mRNA | BC084178.1 | - | 6 - 7 | 570 | 83 | |
More Information
Set Membership: |
|

timm23
Genomic Map