Assay Details
| Gene Symbol: higd1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011058.1 | NP_001011058.1 | 2 - 3 | 152 | 63 | |
| GenBank mRNA | BC084187.1 | - | 2 - 3 | 152 | 63 | |
| CR762016.2 | - | 1 - 2 | 135 | 63 | ||
More Information
Set Membership: |
|

higd1a
Genomic Map