Assay Details
| Gene Symbol: ctsh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011295.1 | NP_001011295.1 | 5 - 6 | 649 | 78 | |
| GenBank mRNA | BC088013.1 | - | 5 - 6 | 649 | 78 | |
More Information
Set Membership: |
|

ctsh
Genomic Map