Assay Details
| Gene Symbol: hsd17b6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011296.1 | NP_001011296.1 | 4 - 5 | 817 | 73 | |
| XM_012956856.2 | XP_012812310.1 | 4 - 5 | 800 | 73 | ||
| GenBank mRNA | BC088014.1 | - | 4 - 5 | 817 | 73 | |
More Information
Set Membership: |
|

hsd17b6
Genomic Map