Assay Details
Gene Symbol: LOC394444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203522.1 | NP_988853.1 | 1 - 2 | 423 | 91 | |
GenBank mRNA | AY079193.1 | - | 1 - 2 | 423 | 91 | |
BC061428.1 | - | 1 - 2 | 405 | 91 | ||
BC081316.1 | - | 1 - 2 | 414 | 91 | ||
BC167263.1 | - | 1 - 2 | 409 | 91 | ||
CR761591.2 | - | 1 - 2 | 413 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
