Assay Details
| Gene Symbol: esr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203535.1 | NP_988866.1 | 3 - 4 | 649 | 64 | |
| GenBank mRNA | AB244211.1 | - | 3 - 4 | 857 | 64 | |
| AY310902.1 | - | 3 - 4 | 649 | 64 | ||
More Information
Set Membership: |
|

esr1
Genomic Map