Assay Details
| Gene Symbol: ergic2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203538.1 | NP_988869.1 | 7 - 8 | 575 | 68 | |
| XM_012953907.1 | XP_012809361.1 | 7 - 8 | 643 | 68 | ||
| XM_018091994.1 | XP_017947483.1 | 7 - 8 | 1296 | 68 | ||
| XM_018091996.1 | XP_017947485.1 | 7 - 8 | 561 | 68 | ||
| GenBank mRNA | BC059763.1 | - | 7 - 8 | 575 | 68 | |
| CR760817.2 | - | 7 - 8 | 591 | 68 | ||
More Information
Set Membership: |
|

ergic2
Genomic Map