Assay Details
| Gene Symbol: cycs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203564.1 | NP_988895.1 | 2 - 3 | 338 | 112 | |
| XM_012964419.2 | XP_012819873.1 | 3 - 4 | 444 | 112 | ||
| GenBank mRNA | BC059740.1 | - | 2 - 3 | 338 | 112 | |
| CR760663.2 | - | 2 - 3 | 194 | 112 | ||
More Information
Set Membership: |
|

cycs
Genomic Map