Assay Details
| Gene Symbol: tmpo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203608.1 | NP_988939.1 | 8 - 9 | 1126 | 71 | |
| XM_012958743.1 | XP_012814197.1 | 11 - 12 | 1641 | 71 | ||
| XM_012958744.1 | XP_012814198.1 | 10 - 11 | 1548 | 71 | ||
| XM_012958745.1 | XP_012814199.1 | 10 - 11 | 1533 | 71 | ||
| XM_012958746.1 | XP_012814200.1 | 9 - 10 | 1428 | 71 | ||
| GenBank mRNA | BC061384.1 | - | 8 - 9 | 1126 | 71 | |
More Information
Set Membership: |
|

tmpo
Genomic Map