Assay Details
| Gene Symbol: sppl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203611.1 | NP_988942.1 | 8 - 9 | 873 | 98 | |
| XM_012958101.2 | XP_012813555.1 | 7 - 8 | 937 | 98 | ||
| GenBank mRNA | BC061375.1 | - | 8 - 9 | 873 | 98 | |
| CR848184.2 | - | 7 - 8 | 741 | 98 | ||
More Information
Set Membership: |
|

sppl3
Genomic Map