Assay Details
| Gene Symbol: ppp4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203612.1 | NP_988943.1 | 2 - 3 | 190 | 71 | |
| XM_018090468.1 | XP_017945957.1 | 2 - 3 | 517 | 71 | ||
| XM_018090469.1 | XP_017945958.1 | 2 - 3 | 207 | 71 | ||
| XM_018090470.1 | XP_017945959.1 | 1 - 2 | 129 | 71 | ||
| GenBank mRNA | BC061369.1 | - | 2 - 3 | 190 | 71 | |
More Information
Set Membership: |
|

ppp4c
Genomic Map