Assay Details
| Gene Symbol: cdc20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203614.1 | NP_988945.1 | 2 - 3 | 288 | 82 | |
| XM_012960717.1 | XP_012816171.1 | 2 - 3 | 333 | 82 | ||
| GenBank mRNA | BC061363.1 | - | 2 - 3 | 288 | 82 | |
| CR760268.2 | - | 1 - 2 | 226 | 82 | ||
More Information
Set Membership: |
|

cdc20
Genomic Map