Assay Details
| Gene Symbol: ube2m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203625.1 | NP_988956.1 | 4 - 5 | 494 | 98 | |
| GenBank mRNA | BC061289.1 | - | 4 - 5 | 494 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ube2m| Gene Symbol: ube2m | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203625.1 | NP_988956.1 | 4 - 5 | 494 | 98 | |
| GenBank mRNA | BC061289.1 | - | 4 - 5 | 494 | 98 | |
Set Membership: |
|