Assay Details
| Gene Symbol: acat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203634.1 | NP_988965.1 | 2 - 3 | 270 | 73 | |
| GenBank mRNA | BC061429.1 | - | 2 - 3 | 270 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

acat2| Gene Symbol: acat2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203634.1 | NP_988965.1 | 2 - 3 | 270 | 73 | |
| GenBank mRNA | BC061429.1 | - | 2 - 3 | 270 | 73 | |
Set Membership: |
|