Assay Details
| Gene Symbol: sec13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203636.1 | NP_988967.1 | 7 - 8 | 919 | 69 | |
| GenBank mRNA | BC061419.1 | - | 7 - 8 | 919 | 69 | |
| CR855621.2 | - | 6 - 7 | 714 | 69 | ||
More Information
Set Membership: |
|

sec13
Genomic Map