Assay Details
| Gene Symbol: lgals3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203655.1 | NP_988986.1 | 4 - 5 | 480 | 64 | |
| GenBank mRNA | BC061319.1 | - | 4 - 5 | 480 | 64 | |
| CR760865.2 | - | 4 - 5 | 478 | 64 | ||
More Information
Set Membership: |
|

lgals3
Genomic Map