Assay Details
| Gene Symbol: rbbp8nl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203678.1 | NP_989009.1 | 8 - 9 | 900 | 63 | |
| GenBank mRNA | BC061355.1 | - | 8 - 9 | 900 | 63 | |
More Information
Set Membership: |
|

rbbp8nl
Genomic Map