Assay Details
| Gene Symbol: rbm24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203685.1 | NP_989016.1 | 2 - 3 | 506 | 95 | |
| GenBank mRNA | BC061322.1 | - | 2 - 3 | 506 | 95 | |
| CT030513.1 | - | 2 - 3 | 515 | 95 | ||
More Information
Set Membership: |
|

rbm24
Genomic Map