Assay Details
| Gene Symbol: mplkip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203694.1 | NP_989025.1 | 1 - 2 | 518 | 72 | |
| GenBank mRNA | BC061411.1 | - | 1 - 2 | 518 | 72 | |
| CT030633.1 | - | 1 - 2 | 523 | 72 | ||
More Information
Set Membership: |
|

mplkip
Genomic Map