Assay Details
| Gene Symbol: gpd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203696.1 | NP_989027.1 | 2 - 3 | 394 | 67 | |
| GenBank mRNA | BC061407.1 | - | 2 - 3 | 394 | 67 | |
| CT030590.1 | - | 2 - 3 | 334 | 67 | ||
More Information
Set Membership: |
|

gpd1l
Genomic Map