Assay Details
Gene Symbol: ube2e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203701.1 | NP_989032.1 | 2 - 3 | 532 | 99 | |
GenBank mRNA | BC061394.1 | - | 2 - 3 | 532 | 99 | |
CR761582.2 | - | 2 - 3 | 233 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
