Assay Details
| Gene Symbol: supt3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203712.1 | NP_989043.1 | 3 - 4 | 267 | 67 | |
| XM_012970255.2 | XP_012825709.1 | 3 - 4 | 374 | 67 | ||
| XM_012970256.2 | XP_012825710.1 | 3 - 4 | 375 | 67 | ||
| XM_018093820.1 | XP_017949309.1 | 3 - 4 | 375 | 67 | ||
| XM_018093821.1 | XP_017949310.1 | 3 - 4 | 375 | 67 | ||
| GenBank mRNA | BC061367.1 | - | 3 - 4 | 267 | 67 | |
| CT030110.1 | - | 3 - 4 | 244 | 67 | ||
More Information
Set Membership: |
|

supt3h
Genomic Map