Assay Details
| Gene Symbol: rrm2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203717.1 | NP_989048.1 | 2 - 3 | 247 | 96 | |
| GenBank mRNA | BC061353.1 | - | 2 - 3 | 247 | 96 | |
More Information
Set Membership: |
|

rrm2.1
Genomic Map