Assay Details
| Gene Symbol: rrm2.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203717.1 | NP_989048.1 | 8 - 9 | 971 | 99 | |
| XM_012962837.2 | XP_012818291.1 | 7 - 8 | 884 | 99 | ||
| GenBank mRNA | BC061353.1 | - | 8 - 9 | 971 | 99 | |
More Information
Set Membership: |
|

rrm2.1
Genomic Map