Assay Details
| Gene Symbol: rbm14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203719.1 | NP_989050.1 | 2 - 3 | 1512 | 92 | |
| GenBank mRNA | BC061344.1 | - | 2 - 3 | 1512 | 92 | |
| CR760510.2 | - | 2 - 3 | 1287 | 92 | ||
More Information
Set Membership: |
|

rbm14
Genomic Map