Assay Details
| Gene Symbol: dazl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203748.1 | NP_989079.1 | 6 - 7 | 534 | 58 | |
| GenBank mRNA | AB109328.1 | - | 6 - 7 | 534 | 58 | |
| BC067947.2 | - | 6 - 7 | 537 | 58 | ||
More Information
Set Membership: |
|

dazl
Genomic Map