Assay Details
| Gene Symbol: serpinf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203755.1 | NP_989086.1 | 6 - 7 | 833 | 70 | |
| XM_018091143.1 | XP_017946632.1 | 5 - 6 | 783 | 70 | ||
| GenBank mRNA | BC062520.1 | - | 6 - 7 | 833 | 70 | |
More Information
Set Membership: |
|

serpinf1
Genomic Map