Assay Details
| Gene Symbol: nt5c1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203765.1 | NP_989096.1 | 4 - 5 | 715 | 87 | |
| GenBank mRNA | BC062488.1 | - | 4 - 5 | 715 | 87 | |
More Information
Set Membership: |
|

nt5c1a
Genomic Map