Assay Details
| Gene Symbol: slc35a3.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203767.1 | NP_989098.1 | 6 - 7 | 845 | 94 | |
| XM_012966293.2 | XP_012821747.1 | 6 - 7 | 906 | 94 | ||
| XM_012966294.2 | XP_012821748.1 | 8 - 9 | 1047 | 94 | ||
| XM_012966295.2 | XP_012821749.1 | 8 - 9 | 1043 | 94 | ||
| XM_012966296.2 | XP_012821750.1 | 8 - 9 | 1033 | 94 | ||
| XM_012966297.2 | XP_012821751.1 | 7 - 8 | 998 | 94 | ||
| XM_012966298.2 | XP_012821752.1 | 7 - 8 | 994 | 94 | ||
| XM_012966299.2 | XP_012821753.1 | 6 - 7 | 1242 | 94 | ||
| GenBank mRNA | BC062483.1 | - | 6 - 7 | 845 | 94 | |
More Information
Set Membership: |
|

slc35a3.2
Genomic Map