Assay Details
| Gene Symbol: naa20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203779.1 | NP_989110.1 | 3 - 4 | 206 | 69 | |
| XM_012962839.2 | XP_012818293.1 | 3 - 4 | 273 | 69 | ||
| GenBank mRNA | BC062502.1 | - | 3 - 4 | 206 | 69 | |
| CT025441.1 | - | 3 - 4 | 199 | 69 | ||
More Information
Set Membership: |
|

naa20
Genomic Map