Assay Details
| Gene Symbol: psma6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203782.1 | NP_989113.1 | 5 - 6 | 609 | 65 | |
| GenBank mRNA | BC061438.1 | - | 5 - 6 | 609 | 65 | |
| CR761245.2 | - | 5 - 6 | 602 | 65 | ||
More Information
Set Membership: |
|

psma6
Genomic Map