Assay Details
| Gene Symbol: tomm40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203794.1 | NP_989125.1 | 4 - 5 | 396 | 75 | |
| XM_012956650.2 | XP_012812104.1 | 4 - 5 | 561 | 75 | ||
| GenBank mRNA | BC061406.1 | - | 4 - 5 | 396 | 75 | |
| CR762023.2 | - | 3 - 4 | 371 | 75 | ||
More Information
Set Membership: |
|

tomm40
Genomic Map