Assay Details
| Gene Symbol: aldob | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203800.1 | NP_989131.1 | 5 - 6 | 664 | 91 | |
| XM_012970260.1 | XP_012825714.1 | 5 - 6 | 988 | 91 | ||
| XM_012970261.2 | XP_012825715.1 | 5 - 6 | 730 | 91 | ||
| GenBank mRNA | BC061442.1 | - | 5 - 6 | 664 | 91 | |
| CR760820.2 | - | 4 - 5 | 625 | 91 | ||
More Information
Set Membership: |
|

aldob
Genomic Map