Assay Details
Gene Symbol: eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203813.1 | NP_989144.1 | 10 - 11 | 1215 | 67 | |
XM_012966301.1 | XP_012821755.1 | 10 - 11 | 1318 | 67 | ||
XM_012966302.1 | XP_012821756.1 | 10 - 11 | 1292 | 67 | ||
XM_012966304.2 | XP_012821758.1 | 10 - 11 | 1288 | 67 | ||
GenBank mRNA | BC061287.1 | - | 10 - 11 | 1215 | 67 | |
BC158432.1 | - | 10 - 11 | 1211 | 67 | ||
CT025514.2 | - | 10 - 11 | 1204 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
