Assay Details
| Gene Symbol: rps6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203821.1 | NP_989152.1 | 2 - 3 | 167 | 97 | |
| GenBank mRNA | BC061628.1 | - | 2 - 3 | 167 | 97 | |
| BC082345.1 | - | 2 - 3 | 160 | 97 | ||
| BC161304.1 | - | 160 | 97 | |||
| CR760876.2 | - | 2 - 3 | 154 | 97 | ||
| Gene Symbol: tmed2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC169186.1 | - | 1793 | 97 | ||
More Information
Set Membership: |
|

rps6
Genomic Map