Assay Details
| Gene Symbol: psmc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203824.1 | NP_989155.1 | 4 - 5 | 301 | 69 | |
| GenBank mRNA | BC061627.1 | - | 4 - 5 | 301 | 69 | |
| BC080137.1 | - | 4 - 5 | 371 | 69 | ||
| CR760112.2 | - | 4 - 5 | 306 | 69 | ||
More Information
Set Membership: |
|

psmc2
Genomic Map