Assay Details
| Gene Symbol: hdgfl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203845.1 | NP_989176.1 | 3 - 4 | 391 | 96 | |
| GenBank mRNA | BC063366.1 | - | 3 - 4 | 391 | 96 | |
| CT025269.2 | - | 3 - 4 | 407 | 96 | ||
More Information
Set Membership: |
|

hdgfl2
Genomic Map