Assay Details
| Gene Symbol: snd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203852.1 | NP_989183.2 | 17 - 18 | 1971 | 61 | |
| GenBank mRNA | BC063211.1 | - | 17 - 18 | 1971 | 61 | |
More Information
Set Membership: |
|

snd1
Genomic Map