Assay Details
| Gene Symbol: txndc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203855.1 | NP_989186.1 | 9 - 10 | 1145 | 56 | |
| GenBank mRNA | BC063355.1 | - | 9 - 10 | 1145 | 56 | |
More Information
Set Membership: |
|

txndc5
Genomic Map