Assay Details
| Gene Symbol: smu1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203857.1 | NP_989188.1 | 11 - 12 | 1510 | 71 | |
| GenBank mRNA | BC063369.1 | - | 11 - 12 | 1510 | 71 | |
| CR855593.2 | - | 11 - 12 | 1463 | 71 | ||
More Information
Set Membership: |
|

smu1
Genomic Map