Assay Details
| Gene Symbol: banp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203864.1 | NP_989195.1 | 12 - 13 | 1576 | 56 | |
| XM_018092972.1 | XP_017948461.1 | 11 - 12 | 1558 | 56 | ||
| GenBank mRNA | BC063204.1 | - | 12 - 13 | 1576 | 56 | |
| BC081348.1 | - | 12 - 13 | 1600 | 56 | ||
| CR848615.2 | - | 12 - 13 | 1553 | 56 | ||
More Information
Set Membership: |
|

banp
Genomic Map