Assay Details
| Gene Symbol: cfap410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203871.1 | NP_989202.1 | 3 - 4 | 396 | 83 | |
| XM_018097068.1 | XP_017952557.1 | 4 - 5 | 477 | 83 | ||
| XM_018097069.1 | XP_017952558.1 | 4 - 5 | 322 | 83 | ||
| GenBank mRNA | BC063358.1 | - | 3 - 4 | 396 | 83 | |
More Information
Set Membership: |
|

cfap410
Genomic Map