Assay Details
| Gene Symbol: rbm4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203873.1 | NP_989204.1 | 3 - 4 | 1119 | 67 | |
| GenBank mRNA | BC063354.1 | - | 3 - 4 | 1119 | 67 | |
| CR942584.2 | - | 3 - 4 | 1069 | 67 | ||
More Information
Set Membership: |
|

rbm4b
Genomic Map