Assay Details
| Gene Symbol: cdx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203878.1 | NP_989209.1 | 2 - 3 | 727 | 65 | |
| GenBank mRNA | AF417197.1 | - | 2 - 3 | 709 | 65 | |
| BC063344.1 | - | 2 - 3 | 727 | 65 | ||
| CR926168.2 | - | 2 - 3 | 708 | 65 | ||
More Information
Set Membership: |
|

cdx2
Genomic Map