Assay Details
| Gene Symbol: klhl41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203883.1 | NP_989214.1 | 5 - 6 | 1774 | 66 | |
| GenBank mRNA | BC063333.1 | - | 5 - 6 | 1774 | 66 | |
More Information
Set Membership: |
|

klhl41
Genomic Map