Assay Details
| Gene Symbol: ubqln1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203890.1 | NP_989221.1 | 1 - 2 | 407 | 65 | |
| GenBank mRNA | BC063190.1 | - | 1 - 2 | 407 | 65 | |
| CT025388.2 | - | 1 - 2 | 369 | 65 | ||
More Information
Set Membership: |
|

ubqln1
Genomic Map