Assay Details
| Gene Symbol: ctsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203894.1 | NP_989225.1 | 6 - 7 | 572 | 92 | |
| XM_012964019.1 | XP_012819473.1 | 6 - 7 | 966 | 92 | ||
| XM_012964020.2 | XP_012819474.1 | 6 - 7 | 571 | 92 | ||
| GenBank mRNA | BC063365.1 | - | 6 - 7 | 572 | 92 | |
More Information
Set Membership: |
|

ctsb
Genomic Map