Assay Details
| Gene Symbol: cdk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203915.1 | NP_989246.1 | 4 - 5 | 556 | 67 | |
| GenBank mRNA | BC063921.1 | - | 4 - 5 | 556 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cdk8| Gene Symbol: cdk8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203915.1 | NP_989246.1 | 4 - 5 | 556 | 67 | |
| GenBank mRNA | BC063921.1 | - | 4 - 5 | 556 | 67 | |
Set Membership: |
|